Learn More
Sino Biological Anti-TIM-1/HAVCR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11051RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HAVCR1 (rh HAVCR1; Catalog#11051-H08H1; AAC39862.1; Met 1-Val 135). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2093