Learn More
Sino Biological Anti-TIM-3/HAVCR2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 110205T04100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminal of the Human TIM-3, and purified by antigen affinity chromatography.
Catalog No. 50-240-9855