Learn More
Sino Biological Recombinant Human TIM-3/HAVCR2 Protein (ECD, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10390H08HB100
Encompass_Preferred
A DNA sequence encoding the human HAVCR2 (AAL65157.1) (Met1-Arg200) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5333