Learn More
Sino Biological Recombinant Human TIM-3/HAVCR2 Protein (mFc Tag), ECD
Supplier:  Sino Biological 10390H38H100
Encompass_Preferred
A DNA sequence encoding the human HAVCR2 (NP_116171.3) (Met1-Arg200) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-5332