Learn More
BIOHIPPO Human HbA1c Microsample
SDP
Catalog No. 5000313981
Encompass
Human HbA1c Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HbA1c in plasma It provides a detection range of 7 81-500 ng/mL with a sensitivity of 2 23 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HbA1c Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HbA1c Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HbA1c biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13981 Supplier BIOHIPPO Supplier No. BHE15211269
Human HbA1c Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HbA1c in plasma It provides a detection range of 7 81-500 ng/mL with a sensitivity of 2 23 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HbA1c Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HbA1c Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HbA1c biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color