Learn More
BIOHIPPO Human HBsAg Microsample
SDP
Catalog No. 5000315721
Encompass
Human HBsAg Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HBsAg in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 31-20 ng/mL with a sensitivity of 0 19 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HBsAg Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HBsAg Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HBsAg biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15721 Supplier BIOHIPPO Supplier No. BHE15210508
Human HBsAg Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HBsAg in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 31-20 ng/mL with a sensitivity of 0 19 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HBsAg Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HBsAg Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HBsAg biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color