Learn More
Proteintech Group Inc Human HDAC1 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0003396T
Encompass_Preferred
Histone deacetylases (HDAC) are a class of enzymes that remove the acetyl groups from the lysine residues leading to the formation of a condensed and transcriptionally silenced chromatin. The protein encoded by this gene belongs to the histone deacetylase/ acuc/ apha family and is a component of the histone deacetylase complex, which is responsible for gene expression silencing. It also plays an important role in the control of cell proliferation and differentiation by interacting with RB, p53 and other transcription factors. HADC inhibition causes apoptosis in tumor cells and HDAC inhibitors may be developed as anti-cancer agents. This kit is used to quantify HDAC1 level in vivo.
Catalog No. 50-197-4009