Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HDAC4 (aa 1071-1084) Synthetic Peptide

Catalog No. PEP141 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP141 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP141 Supplier Invitrogen™ Supplier No. PEP141
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

PEP-141 is a 14 amino acid synthetic peptide whose sequences are derived from amino acids 1071-1084 of human HDAC4. The sequence of this peptide is (amino to carboxy terminus): K (1071)-R-P-D-E-E-P-M-E-E-E-P-P-L (1084) PEP-141 may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human HDAC4, catalog # PA1-863, Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 mL of distilled water.

Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9759
Species Human
Purification Method purified
Accession Number P56524
Common Name HDAC4
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation Peptide with no preservative
For Use With (Application) Control
Gene Alias 4932408F19Rik; AHO3; AI047285; BDMR; DKFZp779K1053; HA6116; HD4; HD7; HD7b; HD9; HDAC; HDAC4; HDAC-4; HDAC7; HDAC7B; HDAC9; HDAC9B; HDAC9FL; HDACA; HDAC-A; HDRP; Histone deacetylase 4; histone deacetylase A; KIAA0288; KIAA0744; MITR
Gene Symbol HDAC4
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Sequence K(1071)-R-P-D-E-E-P-M-E-E-E-P-P-L(1084)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.