Learn More
Sino Biological Anti-HDAC6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105099T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human HDAC6, and purified by antigen affinity chromatography.
Catalog No. 50-240-8176