Learn More
BIOHIPPO Human HDLBP Microsample
SDP
Catalog No. 5000314903
Encompass
Human HDLBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HDLBP in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 128 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HDLBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HDLBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HDLBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14903 Supplier BIOHIPPO Supplier No. BHE15211410
Human HDLBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HDLBP in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 128 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HDLBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HDLBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HDLBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color