Learn More
Zyagen Labs Human Heart cDNA-Random Primer, 30 reactions
SDP
Catalog No. NC1879532
Encompass_Preferred
Human Heart full length tissue cDNA is made of RNA extracted from freshly harvested tissues of single healthy normal adult donor using classical guanidine isothiocyanate phenol: chloroform extraction method. RNA is treated with RNase-free DNase-1 to remove residual DNA. The cDNA is primed with Random Hexamer primer.
Catalog No. NC1879532 Supplier Zyagen Labs Supplier No. HD801HR
Human Heart full length tissue cDNA is made of RNA extracted from freshly harvested tissues of single healthy normal adult donor using classical guanidine isothiocyanate phenol: chloroform extraction method. RNA is treated with RNase-free DNase-1 to remove residual DNA. The cDNA is primed with Random Hexamer primer.