Learn More
American Research Products Inc Human heart fatty acid binding
Catalog No. 5000328743
Encompass
Human heart fatty acid binding protein h-FABP ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28743 Supplier American Research Products Inc Supplier No. CSBE09185H
Human heart fatty acid binding protein h-FABP ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich