Learn More
Biomatik Corporation Human Heat Shock 40kDa Protein 4 (HSPF4) ELISA Kit
Supplier:  Biomatik Corporation EKU12488
Encompass_Preferred
Human Heat Shock 40kDa Protein 4 (HSPF4) ELISA Kit for detecting HSPF4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31689. Gene ID: 3301. Target Synonyms: DNAJA1; DJ-2; DjA1; HDJ2; HSDJ; HSJ2; HSP-F4; hDJ-2; NEDD7; DnaJ (Hsp40) Homolog,Subfamily A,Member 1; DnaJ protein homolog 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7957