Learn More
Biomatik Corporation Human Heat Shock 40kDa Protein 2 (HSPF2) ELISA Kit
Supplier:  Biomatik Corporation EKU04646
Encompass_Preferred
Human Heat Shock 40kDa Protein 2 (HSPF2) ELISA Kit for detecting HSPF2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NNZ3. Gene ID: 3338. Target Synonyms: DNAJC4; MCG18; HSP-F2; DnaJ (Hsp40) Homolog,Subfamily C,Member 4; DnaJ-like protein HSPF2; Multiple endocrine neoplasia type 1 candidate protein number 18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4782