Learn More
Biomatik Corporation Human Heat Shock 70kDa Protein 8 (HSPA8) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12013
Encompass_Preferred
Human Heat Shock 70kDa Protein 8 (HSPA8) High-sensitive ELISA Kit for detecting HSPA8 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11142. Gene ID: 3312. Target Synonyms: LAP1; HSC54; HSC70; HSC71; HSP71; HSPA10; NIP71; HSP73; Heat shock cognate 71 kDa protein; Lipopolysaccharide-associated protein 1; LPS-associated protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7293