Learn More
Biomatik Corporation Human Heat Shock 70kDa Protein 12B (HSPA12B) ELISA Kit
Supplier:  Biomatik Corporation EKU04652
Encompass_Preferred
Human Heat Shock 70kDa Protein 12B (HSPA12B) ELISA Kit for detecting HSPA12B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.128ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96MM6. Gene ID: 116835. Target Synonyms: C20orf60. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8941