Learn More
Biomatik Corporation Human Heat Shock Protein 70 (HSP70) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11940
Encompass_Preferred
Human Heat Shock Protein 70 (HSP70) Mini sample ELISA Kit for detecting HSP70 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.19ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0DMV8. Gene ID: 3303. Target Synonyms: DnaK; 70 Kilodalton Heat Shock Proteins. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5541