Learn More
American Research Products Inc Human heat shock protein 90a
Catalog No. 5000329430
Encompass
Human heat shock protein 90a (HSP-90a) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29430 Supplier American Research Products Inc Supplier No. CSBE13462H
Human heat shock protein 90a (HSP-90a) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich