Learn More
Biomatik Corporation Human Heat Shock Protein Beta 3 (HSPb3) ELISA Kit
Supplier:  Biomatik Corporation EKU04687
Encompass_Preferred
Human Heat Shock Protein Beta 3 (HSPb3) ELISA Kit for detecting HSPb3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12988. Gene ID: 8988. Target Synonyms: HSP-B3; HSPL27; HSP27; HSP 17; Heat Shock 27kDa Protein 3; Heat shock 17 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6313