Learn More
Biomatik Corporation Human Heat Shock Protein Beta 8 (HSPb8) ELISA Kit
Supplier:  Biomatik Corporation EKU04692
Encompass_Preferred
Human Heat Shock Protein Beta 8 (HSPb8) ELISA Kit for detecting HSPb8 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UJY1. Gene ID: 26353. Target Synonyms: CMT2L; DHMN2; E2IG1; H11; HMN2; HSP22; Heat Shock 22kDa Protein 8; Alpha-crystallin C chain; E2-induced gene 1 protein; Small stress protein-like protein HSP22. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8939