Learn More
Biomatik Corporation Human Heat Shock Protein Beta 2 (HSPb2) ELISA Kit
Supplier:  Biomatik Corporation EKU04685
Encompass_Preferred
Human Heat Shock Protein Beta 2 (HSPb2) ELISA Kit for detecting HSPb2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.228ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16082. Gene ID: 3316. Target Synonyms: HSP-B2; MKBP; HSP27; Heat Shock 27kDa Protein 2; DMPK-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8940