Learn More
Biomatik Corporation Human Heat Shock Protein Beta 7 (HSPb7) ELISA Kit
Supplier:  Biomatik Corporation EKU04691
Encompass_Preferred
Human Heat Shock Protein Beta 7 (HSPb7) ELISA Kit for detecting HSPb7 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBY9. Gene ID: 27129. Target Synonyms: HSP-B7; cvHSP; Cardiovascular HSP; Heat Shock 27kDa Protein 7; Cardiovascular heat shock protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6409