Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa) Recombinant Protein, PeproTech®

Catalog No. 01672612
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-612 500 μg
01-672-634 1 mg
01-672-609 100 μg
01-672-615 10 μg
01-672-623 250 μg
01-672-629 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-672-612 Supplier Gibco™ Supplier No. 10018BHS500UG
Only null left
Add to Cart
Edge
Add to Cart

Recombinant Protein

FGF-basic loses its activity over time in solution at physiological temperatures and needs to be re-supplemented periodically to maintain its performance. As a result of selected amino acid substitutions, Heat Stable FGF-basic retains full biological activity and also maintains full activity over extended periods of time in solution at 370 C. These properties make Heat Stable FGF-basic a superior choice for use in cell culture media applications. Recombinant Heat Stable FGF-basic is a 17.2 kDa protein containing 154 amino acids. Human FGF-basic (FGF-2/bFGF) (154 aa), Recombinant Protein PeproTech® was developed under a license from Enantis. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09038
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 2247
Molecular Weight (g/mol) 17.2 kDa
Name Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa)
Purification Method HPLC, SDS-PAGE
Quantity 500 μg
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Basic fibroblast growth factor; basic fibroblast growth factor bFGF; bFGF; FGF; fgf basic; Fgf2; Fgf-2; Fgfb; FGF-b; Fibroblast growth factor; fibroblast growth factor 2; fibroblast growth factor 2 (basic); HBGF-2; Heparin-binding growth factor 2; H-FGF-b-147; H-FGF-b-154; M-FGF-b; Prostatic growth factor; prostatropin
Common Name FGF2
Gene Symbol FGF2
Biological Activity Determined by a cell proliferation assay using Balb/c 3t3 cells. The expected ED50 range is 0.05-0.25 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Thermostable variant of human FGF-basic (154 amino acids) with proprietary amino acid substitutions.
Content And Storage -20°C
Activity Determined by a cell proliferation assay using Balb/c 3t3 cells. The expected ED50 range is 0.05-0.25 ng/ml
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.