Learn More
Biomatik Corporation Human Hedgehog Homolog, Sonic (SHH) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11976
Encompass_Preferred
Human Hedgehog Homolog, Sonic (SHH) Mini sample ELISA Kit for detecting SHH in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15465. Gene ID: 6469. Target Synonyms: HHG1; HLP3; HPE3; SMMCI; SHHN; SHH-N. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4513