Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Hematopoietic Stem Cell Expansion Cytokine Package (IL-3), PeproTech®

Catalog No. HHSC3
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HHSC3 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HHSC3 Supplier Gibco™ Supplier No. HHSC3
  • $2,230.00
    $1,672.65 / Each of 1

    Save $557.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.

Flt3 ligand is a growth factor that regulates proliferation of early hematopoietic cells. By itself it does not stimulate proliferation of early hematopoietic cells, but synergizes with other CSFs and interleukins to induce growth and differentiation.

SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation, and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells.

TPO is a lineage-specific growth factor produced in the liver, kidney, and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes and promotes increased circulating levels of platelets in vivo.

IL-6 is a hematopoietic growth factor that promotes the survival, differentiation, and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil, and mast cell lineages.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Product Type Stem Cell Expansion Cytokine Package
Quantity 1 Kit
Content And Storage 100 μg Human Flt3 Ligand (Cat. No. 300-19-100 UG), 100 μg Human SCF (Cat. No. 300-07-100 UG), 100 μg Human TPO (Cat. No. 300-18-100 UG), 10 μg Human IL-3 (Cat. No. 200-03-10 UG)
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.