Learn More
Biomatik Corporation Human Heme Oxygenase 2, Decycling (HO2) ELISA Kit
Supplier:  Biomatik Corporation EKU04709
Encompass_Preferred
Human Heme Oxygenase 2, Decycling (HO2) ELISA Kit for detecting HO2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30519. Gene ID: 3163. Target Synonyms: HMOX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5529