Learn More
Biomatik Corporation Human Hemicentin 1 (HMCN1) ELISA Kit
Supplier:  Biomatik Corporation EKU11998
Encompass_Preferred
Human Hemicentin 1 (HMCN1) ELISA Kit for detecting HMCN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96RW7. Gene ID: 83872. Target Synonyms: ARMD1; AXPC1; FBLN6; FIBL6; Fibulin 6; Age Related Macular Degeneration 1 (Senile Macular Degeneration). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6044