Learn More
Abcam Human Hemoglobin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1577071X96TESTS
Encompass_Preferred
Human Hemoglobin ELISA Kit is a Sandwich ELISA used to quantify Human Hemoglobin with a sensitivity of 0.8445 ng/ml - Colorimetric ELISA kit - 450 nm readout. Works on any standard plate reader - Wide dynamic range – 6.25 - 200 ng/ml - Cited in over 20 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1530324