Learn More
Abcam Human Hemopexin ELISA Kit (HPX), 1 x 96 Tests
Supplier:  Abcam AB1088591X96TESTS
Encompass_Preferred
Human Hemopexin ELISA Kit (HPX) is a Competitive ELISA kit for the measurement of Human Hemopexin (HPX) in Human in Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1840471