Learn More
Biomatik Corporation Human Hemopexin (HPX) CLIA Kit
Supplier:  Biomatik Corporation EKU08950
Encompass_Preferred
Human Hemopexin (HPX) CLIA Kit for detecting HPX in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02790. Gene ID: 3263. Target Synonyms: Haemopexin; Beta-1B-glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8586