Learn More
Biomatik Corporation Human Hemopoietic Cell Kinase (HCK) ELISA Kit
Supplier:  Biomatik Corporation EKU10896
Encompass_Preferred
Human Hemopoietic Cell Kinase (HCK) ELISA Kit for detecting HCK in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08631. Gene ID: 3055. Target Synonyms: JTK9; Hematopoietic cell kinase; p59-HCK/p60-HCK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5679