Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HENMT1 (aa 1-77) Control Fragment Recombinant Protein

Catalog No. RP94398
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94398 Supplier Invitrogen™ Supplier No. RP94398
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55866. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

HENMT1 (HEN Methyltransferase 1) is a small RNA methyltransferase gene crucial for the stability and functionality of PIWI-interacting RNAs (piRNAs). It is located in human chromosome 1 at the p36.31 locus. The HENMT1 protein catalyzes the 2'-O-methylation at the 3' end of piRNAs in a Mg2+ dependent manner. This modification is essential for piRNA maturation and stability, ensuring the repression of transposable elements (TEs) during spermatogenesis and maintaining genomic integrity in male germ cells. Studies have highlighted the impact of HENMT1 mutations causing male infertility due to disrupted piRNA pathway and improper germ cell gene regulation. The enzymatic activity of HENMT1 is not only significant for male fertility but is also involved in maintaining piRNA stability in female germ cells, although females with HENMT1 mutations show subfertility.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5T8I9
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 113802
Name Human HENMT1 (aa 1-77) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 4921515J06Rik; C1orf59; HEN1; HEN1 methyltransferase homolog 1; HEN1 methyltransferase homolog 1 (Arabidopsis); Henmt1; hua enhancer 1 homolog 1; mHEN1; RGD1306230; Small RNA 2'-O-methyltransferase
Common Name HENMT1
Gene Symbol HENMT1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MEENNLQCSSVVDGNFEEVPRETAIQFKPPLYRQRYQFVKNLVDQHEPKKVADLGCGDTSLLRLLKVNPCIELLVGV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.