Learn More
Biomatik Corporation Human Heparanase 2 (HPA2) ELISA Kit
Supplier:  Biomatik Corporation EKU12022
Encompass_Preferred
Human Heparanase 2 (HPA2) ELISA Kit for detecting HPA2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8WWQ2. Gene ID: 60495. Target Synonyms: HPSE2; HPR2; Inactive heparanase-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8052