Learn More
Biomatik Corporation Human Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) CLIA Kit
Supplier:  Biomatik Corporation EKU09477
Encompass_Preferred
Human Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) CLIA Kit for detecting HBEGF in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.02pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q99075. Gene ID: 1839. Target Synonyms: HB-EGF; DTR; DTS; DTSF; HEGFL; Diphtheria Toxin Receptor; Proheparin-binding EGF-like growth factor; Diphtheria toxin receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6680