Learn More
Biomatik Corporation Human Hepatocyte Nuclear Factor 1 Beta (HNF1b) CLIA Kit
Supplier:  Biomatik Corporation EKU09584
Encompass_Preferred
Human Hepatocyte Nuclear Factor 1 Beta (HNF1b) CLIA Kit for detecting HNF1b in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P35680. Gene ID: 6928. Target Synonyms: FJHN; TCF2; HNF1beta; HNF2; LFB3; LF-B3; MODY5; VHNF1; Transcription Factor 2,Hepatic; Variant Hepatic Nuclear Factor; Homeoprotein LFB3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7439