Learn More
BIOHIPPO Human Hepc Microsample
SDP
Catalog No. 5000314101
Encompass
Human Hepc Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Hepc in serum plasma and other biological fluids It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 4 1 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human Hepc 25 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human Hepc 25 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human Hepc 25 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14101 Supplier BIOHIPPO Supplier No. BHE15212455
Human Hepc Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Hepc in serum plasma and other biological fluids It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 4 1 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human Hepc 25 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human Hepc 25 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human Hepc 25 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color