Learn More
Biomatik Corporation Human Hepcidin (Hepc) CLIA Kit
Supplier:  Biomatik Corporation EKU09966
Encompass_Preferred
Human Hepcidin (Hepc) CLIA Kit for detecting Hepc in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 5.49-4,000pg/ml. Sensitivity: 1.89pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P81172. Gene ID: 57817. Target Synonyms: HAMP; HFE2B; PLTR; LEAP1; Hepcidin Antimicrobial Peptide; Liver-expressed antimicrobial peptide 1; Putative liver tumor regressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4694