Learn More
Biomatik Corporation Human Hephaestin (HEPH) ELISA Kit
Supplier:  Biomatik Corporation EKU04767
Encompass_Preferred
Human Hephaestin (HEPH) ELISA Kit for detecting HEPH in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BQS7. Gene ID: 9843. Target Synonyms: CPL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6648