Learn More
Lonza Walkersville Human Heps Single Donor
Catalog No. 5000496309
Encompass
Cryopreserved single donor human hepatocytes are characterized for high viability and general Phase I and Phase II metabolism activity in short term suspension metabolism assays The suspension qualified hepatocytes are ideally suited from short-term ( 4 hrs) in vitro drug clearance assays The suspension qualified hepatocytes are not suitable for applications that require hepatocytes cultured in adhered monolayers
Catalog No. 50-004-96309 Supplier Lonza Walkersville Supplier No. HUCSD
Cryopreserved single donor human hepatocytes are characterized for high viability and general Phase I and Phase II metabolism activity in short term suspension metabolism assays The suspension qualified hepatocytes are ideally suited from short-term ( 4 hrs) in vitro drug clearance assays The suspension qualified hepatocytes are not suitable for applications that require hepatocytes cultured in adhered monolayers