Learn More
ACROBiosystems Alexa Fluor 488-Labeled Human Her2 / ErbB2 Protein, His TagStar Staining
Catalog No. 502332057
Encompass_Preferred
Alexa Fluor 488-Labeled Human Her2, His Tag (HE2-HA2H3) is produced via conjugation of AF488 to Human Her2, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Her2, His Tag is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1).
Catalog No. 50-233-2057 Supplier ACROBiosystems Supplier No. HE2HA2H325UG
Alexa Fluor 488-Labeled Human Her2, His Tag (HE2-HA2H3) is produced via conjugation of AF488 to Human Her2, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Her2, His Tag is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1).