Learn More
ACROBiosystems Human Her2 / ErbB2 (23-510) Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems HE2H52H9100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Her2 (23-510), His Tag (HE2-H52H9) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Ala 510 (Accession # P04626-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.6 kDa. The protein migrates as 65-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Her2 (23-510), His Tag (Cat. No. HE2-H52H9) at 1 μg/mL (100 μL/well) can bind Pertuzumab with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Human Her2 (23-510), His Tag (Cat. No. HE2-H52H9) at 1 μg/mL (100 μL/well) can bind Bispecific Antibody (Her2 × Her3) with a linear range of 0.2-3 ng/mL (Routinely tested).
Catalog No. 50-205-4834