Learn More
Sino Biological Human Her2/ERBB2 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786727
Encompass_Preferred
The Human HER2 / ErbB2 / CD340 ELISA Pair Set is for the quantitative determination of Human HER2 / ErbB2 / CD340.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human HER2 / ErbB2 / CD340 coated on a 96-well plate. Standards and samples are added to the wells, and any Human HER2 / ErbB2 / CD340 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human HER2 / ErbB2 / CD340 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human HER2 / ErbB2 / CD340 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86727 Supplier Sino Biological Supplier No. SEKA1000415
The Human HER2 / ErbB2 / CD340 ELISA Pair Set is for the quantitative determination of Human HER2 / ErbB2 / CD340.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human HER2 / ErbB2 / CD340 coated on a 96-well plate. Standards and samples are added to the wells, and any Human HER2 / ErbB2 / CD340 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human HER2 / ErbB2 / CD340 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human HER2 / ErbB2 / CD340 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.