Learn More
ACROBiosystems Human Her2 / ErbB2 (S310Y) Protein, His Tag (MALS verified)
Catalog No. 502055088
Encompass_Preferred
Recombinant Protein;1MG;Human Her2 (S310Y), His Tag (HE2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1 (S310Y)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 71.3 kDa. The protein migrates as 80-100 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5088 Supplier ACROBiosystems Supplier No. HE2H52H31MG
Recombinant Protein;1MG;Human Her2 (S310Y), His Tag (HE2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1 (S310Y)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 71.3 kDa. The protein migrates as 80-100 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.