Learn More
Biomatik Corporation Human Hermansky Pudlak Syndrome Protein 6 (HPS6) ELISA Kit
Supplier:  Biomatik Corporation EKU04768
Encompass_Preferred
Human Hermansky Pudlak Syndrome Protein 6 (HPS6) ELISA Kit for detecting HPS6 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86YV9. Target Synonyms: Ru; Ruby-eye protein homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7276