Learn More
ATCC Human herpesvirus 1 DNA; HSV-1 strain KOS
Catalog No. 502382910
Encompass
Viral nucleic acids, including virus genomic DNA, in total DNA isolated from Vero cells (ATCC CCL-81) infected with Human herpesvirus 1 (HSV-1) strain KOS (ATCC VR-1493). Virus and host cells are also available through the ATCC Catalog. This product was prepared using methods known to inactivate viruses. It can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2910 Supplier ATCC Supplier No. VR1493D
Viral nucleic acids, including virus genomic DNA, in total DNA isolated from Vero cells (ATCC CCL-81) infected with Human herpesvirus 1 (HSV-1) strain KOS (ATCC VR-1493). Virus and host cells are also available through the ATCC Catalog. This product was prepared using methods known to inactivate viruses. It can be used in PCR and other molecular biology applications.