Learn More
ATCC Human herpesvirus 1; Herpes simplex virus 1; Strain: KOS
Supplier:  ATCC VR1493
Encompass
Human herpesvirus 1 strain KOS is propagated in Vero cells (ATCC CCL-81). This strain was isolated from the lip lesion of a patient with a cold sore and was deposited by the University of Pennsylvania. It has applications in respiratory disease research.
Catalog No. 50-238-4844