Learn More
ATCC Human herpesvirus 1; Herpes simplex virus 1; Strain: HF
Catalog No. 502382965
Encompass
Human herpesvirus 1 strain HF is propagated in Vero cells (ATCC CCL-81). This strain was isolated in 1922 from the vesicle fluid of a human patient with recurrent herpes simplex in New York, US. It has applications in testing disinfectants and infectious disease and sexually transmitted disease research. Vials of this product contain total fluid and material from cell culture infected by virus; cellular debris and possibly cells containing cell-associated virus will be present.
Catalog No. 50-238-2965 Supplier ATCC Supplier No. VR260
Human herpesvirus 1 strain HF is propagated in Vero cells (ATCC CCL-81). This strain was isolated in 1922 from the vesicle fluid of a human patient with recurrent herpes simplex in New York, US. It has applications in testing disinfectants and infectious disease and sexually transmitted disease research. Vials of this product contain total fluid and material from cell culture infected by virus; cellular debris and possibly cells containing cell-associated virus will be present.