Learn More
ATCC Human herpesvirus 1; HSV-1 strain MacIntyre; Quantitative DNA
Catalog No. 502383447
Encompass
Quantitative Genomic DNA from Human herpesvirus 1 strain MacIntyre can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-3447 Supplier ATCC Supplier No. VR539DQ
Quantitative Genomic DNA from Human herpesvirus 1 strain MacIntyre can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.