Learn More
ATCC Human herpesvirus 1; HSV-1 strain MacIntyre; DNA
Catalog No. 502384296
Encompass
Genomic DNA isolated from a preparation of cell lysate and supernatant from Vero cells (ATCC CCL-81) infected with Human herpesvirus 1 (HSV-1) strain MacIntyre (ATCC VR-539). This product was prepared using methods known to inactivate viruses. The product has applications in PCR or other molecular virology procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-4296 Supplier ATCC Supplier No. VR539D
Genomic DNA isolated from a preparation of cell lysate and supernatant from Vero cells (ATCC CCL-81) infected with Human herpesvirus 1 (HSV-1) strain MacIntyre (ATCC VR-539). This product was prepared using methods known to inactivate viruses. The product has applications in PCR or other molecular virology procedures. The source organism and host cells are also available through the ATCC catalog.