Learn More
ATCC Human herpesvirus 1; Strain: F
Catalog No. 502382644
Encompass
Human herpesvirus 1 strain F is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated from the facial vesicle of a human patient and has applications in susceptibility testing and virucide testing.
Catalog No. 50-238-2644 Supplier ATCC Supplier No. VR733
Human herpesvirus 1 strain F is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated from the facial vesicle of a human patient and has applications in susceptibility testing and virucide testing.